missing translation for 'onlineSavingsMsg'
Learn More

Aminoferrocene 96.0+%, TCI America™

Catalog No. A16011G
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A16011G 1 g
A1601100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A16011G Supplier TCI America Supplier No. A16011G
Only null left

Chemical Identifiers

CAS 1273-82-1
Molecular Formula C10H11FeN
MDL Number MFCD03844760
Synonym Ferrocenylamine

Specifications

Quantity 1 g
Formula Weight 201.05
Percent Purity ≥96.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material Aminoferrocene
TSCA No
Recommended Storage Refrigerator