Learn More
(Aminoethylaminomethyl)phenethyltrimethoxysilane, mixture of m and p isomers
As a crosslinking agent | CAS: 74113-77-2 | H2NC2H4NHCH2C6H4C2H4Si(OCH3)3
$62.67 - $199.48
Chemical Identifiers
| CAS | 74113-77-2 |
|---|---|
| Molecular Formula | H2NC2H4NHCH2C6H4C2H4Si(OCH3)3 |
| MDL Number | MFCD00054775 |
| Synonym | aminoethy-laminomethyl phenethyl-trimethoxysilane, 4-2-aminoethylaminomethyl phenethyltrimethoxysilane, n1-4-2-trimethoxysilyl ethyl benzyl ethane-1,2-diamine, n'-4-2-trimethoxysilylethyl phenyl methyl ethane-1,2-diamine, 2-aminoethyl 4-2-trimethoxysilyl ethyl phenyl methyl amine |
Description
As a crosslinking agent
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 74113-77-2 | |
| H2NC2H4NHCH2C6H4C2H4Si(OCH3)3 | |
| MFCD00054775 | |
| aminoethy-laminomethyl phenethyl-trimethoxysilane, 4-2-aminoethylaminomethyl phenethyltrimethoxysilane, n1-4-2-trimethoxysilyl ethyl benzyl ethane-1,2-diamine, n'-4-2-trimethoxysilylethyl phenyl methyl ethane-1,2-diamine, 2-aminoethyl 4-2-trimethoxysilyl ethyl phenyl methyl amine |
Specifications
| 128°C (0.2mmHg) | |
| 74113-77-2 | |
| 2 g | |
| MFCD00054775 | |
| >110°C (230°F) | |
| 298.46 | |
| 1.02 g/mL |
| Colorless to Yellow | |
| Liquid | |
| H2NC2H4NHCH2C6H4C2H4Si(OCH3)3 | |
| UN2735 | |
| aminoethy-laminomethyl phenethyl-trimethoxysilane, 4-2-aminoethylaminomethyl phenethyltrimethoxysilane, n1-4-2-trimethoxysilyl ethyl benzyl ethane-1,2-diamine, n'-4-2-trimethoxysilylethyl phenyl methyl ethane-1,2-diamine, 2-aminoethyl 4-2-trimethoxysilyl ethyl phenyl methyl amine | |
| Air and moisture sensitive | |
| (Aminoethylaminomethyl)phenethyltrimethoxysilane, Mixture of m and p isomers |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
TSCA : Yes
Recommended Storage : Ambient temperatures; Store under Argon
RUO – Research Use Only