missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Aminoallyl-dUTP Solution (10 mM)

Catalog No. FERR0091
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FERR0091 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FERR0091 Supplier Thermo Scientific™ Supplier No. R0091

Thermo Scientific Aminoallyl-dUTP (5-[3-aminoallyl]-2'-deoxyuridine-5'-triphosphate) is supplied as 10 mM and 50 mM aqueous solutions titrated to pH 7.0 with NaOH.

Thermo Scientific Aminoallyl-dUTP (5-[3-aminoallyl]-2'-deoxyuridine-5'-triphosphate) is supplied as 10 mM and 50 mM aqueous solutions titrated to pH 7.0 with NaOH.

Aminoallyl-dUTP can be enzymatically incorporated into DNA with:
• Reverse transcriptases
Taq DNA polymerase (Cat. No. EP0072)
• phi29 DNA Polymerase (EP0091)
• Klenow Fragment (EP0051)
• Klenow Fragment, exo- (EP0421)
• DNA Polymerase I (EP0041)

The resulting amine-containing DNA can be subsequently labeled with any amine-reactive fluorescent dye, biotin, or hapten.

Applications
• Enzymatic indirect non-radioactive labeling of DNA during cDNA synthesis
• PCR
• Nick-translation
• Random-primed labeling
• Primer extension

Specifications

Concentration 10 mM
Content And Storage Aminoallyl-dUTP Solution, 10 mM (100 μL)

Store at –20°C.
Form Liquid
Polymerase DNA Polymerase
Labeling Method Direct Labeling
Label or Dye Aminoallyl
Quantity 100 μL
Shipping Condition Dry Ice
For Use With (Application) cDNA Synthesis, Non-Radioactive Labeling

For Research Use Only. Not for use in diagnostic procedures.