missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aminoacetonitrile Sulfate 98.0+%, TCI America™
$72.70 - $600.22
Chemical Identifiers
| CAS | 5466-22-8 |
|---|---|
| Molecular Formula | C2H4N2 |
| MDL Number | MFCD00012851 |
| Synonym | Cyanomethylamine Sulfate |
Chemical Identifiers
| 5466-22-8 | |
| C2H4N2 | |
| MFCD00012851 | |
| Cyanomethylamine Sulfate |
Specifications
| 5466-22-8 | |
| White | |
| MFCD00012851 | |
| 3439 | |
| 210.21 | |
| Crystalline Powder |
| 165°C | |
| C2H4N2 | |
| 25 g | |
| Cyanomethylamine Sulfate | |
| ≥98.0% (N,T) | |
| Aminoacetonitrile Sulfate |
Safety and Handling
EINECSNumber : (2)-1519
RTECSNumber : MC2100000
TSCA : No