missing translation for 'onlineSavingsMsg'
Learn More

Aminoacetonitrile sulfate, 97%

CAS: 5466-22-8 | C4H8N8·H2SO4

$89.51 - $656.70

Chemical Identifiers

CAS 5466-22-8
Molecular Formula C4H8N8·H2SO4
MDL Number MFCD00012851
Synonym Glycinonitrile sulfate
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB2215514
View Documents
Thermo Scientific Chemicals
B2215514
25 g
Each for $89.51
Only null left
 
AAB2215522
View Documents
Thermo Scientific Chemicals
B2215522
100 g
Each for $233.01
Only null left
 
AAB2215536
View Documents
Thermo Scientific Chemicals
B2215536
500 g
Each for $656.70
Only null left
 
Description

Description

Aminoacetonitrile sulfate is used in organic synthesis, catalytic agent, petrochemical additive.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Aminoacetonitrile sulfate is used in organic synthesis, catalytic agent, petrochemical additive.

Solubility
Soluble in water.

Notes
Hygroscopic. Store away from strong oxidizing agents, water/ moisture.
Specifications

Chemical Identifiers

5466-22-8
C4H8N8·H2SO4
MFCD00012851
Glycinonitrile sulfate

Specifications

5466-22-8
Odorless
(H2NCH2CN)2·H2SO4
25 g
3916094
14,412
Soluble in water.
97%
∼160°C (decomposition)
C4H8N8·H2SO4
MFCD00012851
UN3439
Hygroscopic
Glycinonitrile sulfate
210.21
Aminoacetonitrile sulfate
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H301-H315-H319-H335

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.

EINECSNumber : 226-772-0

RTECSNumber : MC2100000

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only