missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aminoacetaldehyde Dimethyl Acetal 98.0+%, TCI America™
$69.31 - $529.43
Chemical Identifiers
| CAS | 22483-09-6 |
|---|---|
| Molecular Formula | C4H11NO2 |
| Molecular Weight (g/mol) | 105.137 |
| MDL Number | MFCD00008135 |
| InChI Key | QKWWDTYDYOFRJL-UHFFFAOYSA-N |
| Synonym | aminoacetaldehyde dimethyl acetal, 2,2-dimethoxyethylamine, ethanamine, 2,2-dimethoxy, 2,2-dimethoxyethan-1-amine, dimethylaminoacetal, amino acetaldehyde dimethyl acetal, 1-amino-2,2-dimethoxyethane, 2-aminoacetaldehyde dimethyl acetal, acetaldehyde, amino-, dimethyl acetal, acmc-209fwr |
| PubChem CID | 89728 |
| IUPAC Name | 2,2-dimethoxyethanamine |
| SMILES | COC(CN)OC |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
A080125ML
|
TCI America
A080125ML |
25 mL |
Each for $69.31
|
|
|||||
|
A0801250ML
|
TCI America
A0801250ML |
250 mL |
Each for $529.43
|
|
|||||
Chemical Identifiers
| 22483-09-6 | |
| 105.137 | |
| QKWWDTYDYOFRJL-UHFFFAOYSA-N | |
| 89728 | |
| COC(CN)OC |
| C4H11NO2 | |
| MFCD00008135 | |
| aminoacetaldehyde dimethyl acetal, 2,2-dimethoxyethylamine, ethanamine, 2,2-dimethoxy, 2,2-dimethoxyethan-1-amine, dimethylaminoacetal, amino acetaldehyde dimethyl acetal, 1-amino-2,2-dimethoxyethane, 2-aminoacetaldehyde dimethyl acetal, acetaldehyde, amino-, dimethyl acetal, acmc-209fwr | |
| 2,2-dimethoxyethanamine |
Specifications
| 22483-09-6 | |
| 136°C | |
| MFCD00008135 | |
| 2924 | |
| QKWWDTYDYOFRJL-UHFFFAOYSA-N | |
| 2,2-dimethoxyethanamine | |
| 89728 | |
| ≥98.0% (GC,T) | |
| Aminoacetaldehyde Dimethyl Acetal |
| Colorless | |
| C4H11NO2 | |
| 25 mL | |
| aminoacetaldehyde dimethyl acetal, 2,2-dimethoxyethylamine, ethanamine, 2,2-dimethoxy, 2,2-dimethoxyethan-1-amine, dimethylaminoacetal, amino acetaldehyde dimethyl acetal, 1-amino-2,2-dimethoxyethane, 2-aminoacetaldehyde dimethyl acetal, acetaldehyde, amino-, dimethyl acetal, acmc-209fwr | |
| COC(CN)OC | |
| 105.137 | |
| 105.14 | |
| Liquid |
Safety and Handling
TSCA : Yes