Learn More
Aminoacetaldehyde dimethyl acetal, 99%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 153340025
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
| Aminoacetaldehyde dimethyl acetal | |
| 22483-09-6 | |
| 135°C to 139°C (95.0mmHg) | |
| Authentic | |
| Glass bottle | |
| 1.4160 to 1.418 | |
| MFCD00008135 | |
| 04, II, 758 | |
| aminoacetaldehyde dimethyl acetal, 2,2-dimethoxyethylamine, ethanamine, 2,2-dimethoxy, 2,2-dimethoxyethan-1-amine, dimethylaminoacetal, amino acetaldehyde dimethyl acetal, 1-amino-2,2-dimethoxyethane, 2-aminoacetaldehyde dimethyl acetal, acetaldehyde, amino-, dimethyl acetal, acmc-209fwr | |
| QKWWDTYDYOFRJL-UHFFFAOYSA-N | |
| 2,2-dimethoxyethanamine | |
| 89728 | |
| 105.14 |
| 99% | |
| 0.9700g/mL | |
| 41°C | |
| 98.5% min. (GC) | |
| C4H11NO2 | |
| H2NCH2CH(OCH3)2 | |
| 2.5kg | |
| 0.97 | |
| Solubility in water: miscible | |
| COC(CN)OC | |
| 105.14 | |
| 0.15 Pas (20°C) | |
| 99% |
Safety and Handling
GHS H Statement
Flammable liquid and vapor.
Causes severe skin burns and eye damage.
GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi
GHS Signal Word: Danger
EINECSNumber : 245-026-5
TSCA : TSCA
RUO â Research Use Only