Learn More
Aminoacetaldehyde dimethyl acetal, 99%
CAS: 22483-09-6 | C4H11NO2 | 105.137 g/mol
Supplier: Thermo Scientific Chemicals A1549822
Description
Amino acetaldehyde dimethyl was used in preparation of chitosan- dendrimer hybrids having various functional groups such as carboxyl, ester and poly(ethylene glycol). It was used in an synthesis of a bicyclic proline analog from L-ascorbic acid and in 3-component reaction catalyzed by MgClO4 leading to α-aminophosphonates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsAmino acetaldehyde dimethyl was used in preparation of chitosan- dendrimer hybrids having various functional groups such as carboxyl, ester and poly(ethylene glycol). It was used in an synthesis of a bicyclic proline analog from L-ascorbic acid and in 3-component reaction catalyzed by MgClO4 leading to α-aminophosphonates.
Solubility
Miscible in water. Soluble in chloroform, methanol.
Notes
Store in a cool, dry conditions in a well sealed container. Incompatible with oxidizing agent.
Specifications
| Aminoacetaldehyde dimethyl acetal | |
| 0.973 | |
| 41°C (106°F) | |
| 1.417 | |
| 100 g | |
| 741868 | |
| Miscible in water. Soluble in chloroform,methanol. | |
| COC(CN)OC | |
| 105.137 | |
| 105.14 |
| 22483-09-6 | |
| 135°C to 138°C | |
| C4H11NO2 | |
| MFCD00008135 | |
| UN2733 | |
| aminoacetaldehyde dimethyl acetal, 2,2-dimethoxyethylamine, ethanamine, 2,2-dimethoxy, 2,2-dimethoxyethan-1-amine, dimethylaminoacetal, amino acetaldehyde dimethyl acetal, 1-amino-2,2-dimethoxyethane, 2-aminoacetaldehyde dimethyl acetal, acetaldehyde, amino-, dimethyl acetal, acmc-209fwr | |
| QKWWDTYDYOFRJL-UHFFFAOYSA-N | |
| 2,2-dimethoxyethanamine | |
| 89728 | |
| 99% |
Chemical Identifiers
| 22483-09-6 | |
| 105.137 | |
| QKWWDTYDYOFRJL-UHFFFAOYSA-N | |
| 89728 | |
| COC(CN)OC |
| C4H11NO2 | |
| MFCD00008135 | |
| aminoacetaldehyde dimethyl acetal, 2,2-dimethoxyethylamine, ethanamine, 2,2-dimethoxy, 2,2-dimethoxyethan-1-amine, dimethylaminoacetal, amino acetaldehyde dimethyl acetal, 1-amino-2,2-dimethoxyethane, 2-aminoacetaldehyde dimethyl acetal, acetaldehyde, amino-, dimethyl acetal, acmc-209fwr | |
| 2,2-dimethoxyethanamine |
Safety and Handling
GHS H Statement
H314-H318-H226
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapour.
P210-P233-P240-P241-P242-P243-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
H226-H314
DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: AMINES, FLAMMABLE, CORROSIVE, N.O.S.
EINECSNumber : 245-026-5
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only