Learn More
Cytiva Amine Coupling Kit, Reagents for Covalent Immobilization of Molecules
Catalog No. 501055278
Encompass
Reagents for covalent immobilization of molecules carrying a primary amine group.
Catalog No. 501055278 Supplier Cytiva Supplier No. BR100050
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Reagents for covalent immobilization of molecules carrying a primary amine group.