missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Amiloride Hydrochloride Hydrate 98.0+%, TCI America™
Supplier: TCI America A25995G
Specifications
| Amiloride Hydrochloride Hydrate | |
| 299°C | |
| MFCD00211292,MFCD03703482 | |
| amiloride hydrochloride, amiloride hcl, amiloride chloride, amiloride hydrochloride anhydrous, ccris 5959, amilorida hydrochloride, amiloride, hydrochloride, n-amidino-3,5-diamino-6-chloropyrazinecarboxamide hydrochloride, n-amidino-3,5-diamino-6-chloropyrazinecarboxamide monohydrochloride | |
| [H+].[Cl-].NC(N)=NC(=O)C1=NC(Cl)=C(N)N=C1N | |
| 266.09 | |
| CHEBI:84743 | |
| ≥98.0% (HPLC,N) |
| 2016-88-8 | |
| C6H9Cl2N7O | |
| 5 g | |
| ACHKKGDWZVCSNH-UHFFFAOYSA-N | |
| hydrogen 3,5-diamino-6-chloro-N-(diaminomethylidene)pyrazine-2-carboxamide chloride | |
| 16230 | |
| 266.09 | |
| Crystalline Powder |
Chemical Identifiers
| 2016-88-8 | |
| 266.09 | |
| ACHKKGDWZVCSNH-UHFFFAOYSA-N | |
| 16230 | |
| hydrogen 3,5-diamino-6-chloro-N-(diaminomethylidene)pyrazine-2-carboxamide chloride |
| C6H9Cl2N7O | |
| MFCD00211292,MFCD03703482 | |
| amiloride hydrochloride, amiloride hcl, amiloride chloride, amiloride hydrochloride anhydrous, ccris 5959, amilorida hydrochloride, amiloride, hydrochloride, n-amidino-3,5-diamino-6-chloropyrazinecarboxamide hydrochloride, n-amidino-3,5-diamino-6-chloropyrazinecarboxamide monohydrochloride | |
| CHEBI:84743 | |
| [H+].[Cl-].NC(N)=NC(=O)C1=NC(Cl)=C(N)N=C1N |
Safety and Handling
RTECSNumber : UQ2275500
TSCA : No