missing translation for 'onlineSavingsMsg'
Learn More

EVOS™ EVOS™ Vessel Holder, SPL T-75 flask

Catalog No. 12563492
Change view
Click to view available options
Quantity:
1 Holder
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
12563492 1 Holder
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 12563492 Supplier EVOS™ Supplier No. AMEPVH012

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Securely holds one SPL T-75 flask

  • For use with EVOS and Micromaster inverted microscopes
  • Holds one SPL T-75 flask

Specifications

Type Vessel Holder
Product Line EVOS
For Use With (Equipment) EVOS™ XL Core Imaging System, EVOS™ FL Color Imaging System, EVOS™ XL Imaging System, EVOS™ FL Imaging System, EVOS™ FL Auto Imaging System
Format one, SPL T-75 flask
Holds 1 SPL T-75 Flask
Quantity 1 Holder

For Research Use Only. Not for use in diagnostic procedures.