Learn More
AMG Calibration Fluid
Catalog No. 1256043
Change view
Click to view available options
Scale Range:
60 Brix
Catalog No. Scale Range
1256043 60 Brix
1 options
Catalog No. 1256043 Supplier Laxco Inc Supplier No. RP CAL60
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Calibration solution for RHB Model refractometers
  • 46 brix: For use with refractometer Models RHB-62, RHB-82
  • 60 brix: For use with refractometer Model RHB-90
Specifications
Type Calibration Fluid
Scale Range 60 Brix
For Use With (Equipment) RHB-90 Refractometer