missing translation for 'onlineSavingsMsg'
Learn More

Amezinium Methyl Sulfate 98.0+%, TCI America™

$67.11 - $328.08

Chemical Identifiers

CAS 30578-37-1
Molecular Formula C12H15N3O5S
Molecular Weight (g/mol) 313.33
MDL Number MFCD00865972
InChI Key ZEASXVYVFFXULL-UHFFFAOYSA-N
Synonym 4-Amino-6-methoxy-1-phenylpyridazinium Methyl Sulfate
PubChem CID 71926
IUPAC Name 4-amino-6-methoxy-1-phenyl-1λ⁵-pyridazin-1-ylium methyl sulfate
SMILES COS([O-])(=O)=O.COC1=CC(N)=CN=[N+]1C1=CC=CC=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
A24251G
View Documents
TCI America
A24251G
1 g
Each for $67.11
Only null left
 
A24255G
View Documents
TCI America
A24255G
5 g
Each for $328.08
Only null left
 
Description

Description

Specifications

Chemical Identifiers

30578-37-1
313.33
ZEASXVYVFFXULL-UHFFFAOYSA-N
71926
COS([O-])(=O)=O.COC1=CC(N)=CN=[N+]1C1=CC=CC=C1
C12H15N3O5S
MFCD00865972
4-Amino-6-methoxy-1-phenylpyridazinium Methyl Sulfate
4-amino-6-methoxy-1-phenyl-1λ⁵-pyridazin-1-ylium methyl sulfate

Specifications

30578-37-1
White-Yellow
MFCD00865972
4-Amino-6-methoxy-1-phenylpyridazinium Methyl Sulfate
COS([O-])(=O)=O.COC1=CC(N)=CN=[N+]1C1=CC=CC=C1
313.33
313.33
Crystalline Powder
176°C
C12H15N3O5S
1 g
ZEASXVYVFFXULL-UHFFFAOYSA-N
4-amino-6-methoxy-1-phenyl-1λ⁵-pyridazin-1-ylium methyl sulfate
71926
≥98.0% (HPLC)
Amezinium Methyl Sulfate
Videos
Safety and Handling

Safety and Handling

RTECSNumber : UR6066500

TSCA : No

SDS
missing translation for 'documents'

missing translation for 'documents'