missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Amezinium Methyl Sulfate 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 176°C |
| Color | White-Yellow |
| Quantity | 1 g |
| Formula Weight | 313.33 |
| Percent Purity | ≥98.0% (HPLC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Amezinium Methyl Sulfate |
Safety and Handling
| RTECSNumber | UR6066500 |
| TSCA | No |