missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Amersham™ Hyperfilm™ ECL
Description
- Antistatic coating reduces risk of visible artifacts
- Clear base offers high contrast for easy reading of images
- Suitable for use with all blue and green light chemiluminescent systems
- Film can be processed in automatic processors or manually using most common X-ray film developers and fixers
- Chemiluminescent detection in Western, Southern, and Northern blots
Specifications
Specifications
| Product Type | Amersham Hyperfilm ECL |
| For Use With (Application) | Chemiluminescent detection in Western, Southern, and Northern blots. |
| Quantity | 100 Sheets/Box |
| Content And Storage | 4°C |
| Dimensions (LxW) | 20.3 x 25.4 cm (8 x 10 in.) |