missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Amersham™ ECL Plex™ Western Blotting Combination Packs
Complete system optimized for fluorescent Western blotting
$372.00 - $377.00
Specifications
| Product Type | Western Blot Kit |
|---|---|
| For Use With (Application) | ECL Plex western blotting system |
| Storage Requirements | Varies by component |
Includes: Amersham ECL Plex goat-α-mouse IgG-Cy3, 150μg; Amersham ECL Plex goat-α-rabbit IgG-Cy5, 150μg; Amersham ECL Plex Fluorescent Rainbow Markers, 120μL; 45-001-214: Amersham Protran Premium, 10 × 10cm, 10 sheets or 45-001-215: Hybond LFP 0.2μM PVDF, 20 × 20cm, 3 sheets
| Catalog Number | Mfr. No. | Formulation | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Formulation | Price | Quantity | |||||
|
45-001-214
|
Cytiva
RPN998 |
Cy3, Cy5, Amersham Protran Premium 0.45 NC |
Each for $372.00
|
|
|||||
|
45-001-215
|
Cytiva
RPN999 |
Cy3, Cy5, Amersham Hybond LFP 0.2 PVDF |
Each for $377.00
|
|
|||||
Description
- Improved formulation of Cy3 conjugates for even lower background
Specifications
| Western Blot Kit | |
| ECL Plex western blotting system | |
| Varies by component |