missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Amersham™ ECL™ Plex Fluorescent Rainbow Markers
Description
- Monitor progress of protein electrophoresis and assess transfer efficiency and molecular weight of blotted proteins without staining
- Enhanced to enable faster and simpler identification of proteins on SDS-polyacrylamide gels
- Ready-to-load
- Provide sharper, more intense bands on gels and blots and have improved band spacing for more accurate molecular weight determination
- Provides fluorescent images using Cy3 and Cy5 channels
Order Info
Each 120μL pack size is sufficient for use with at least 40 minigels (10 x 8cm) or 20 large gels (20 x 20cm). Each 500μL pack size is sufficient for use with at least 160 minigels (10 x 8cm) or 80 large gels (20 x 20cm).
Specifications
Specifications
| Description | These fluorescent rainbow markers are for use with the ECL Plex Western blotting detection system. |
| Color | Multi |
| Formulation | Supplied in 35% Glycerol and sample buffer containing Mercaptoethanesulphonic Acid (MESNA) as reducing agent |
| For Use With (Application) | For quantitative western blots |
| Quantity | 120 μL |
| Storage Requirements | -20°C |
| Sufficient For | 12 Minigel Loadings |
| Product Type | Protein Molecular Weight Marker |