missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Amersham™ ECL™ DualVue Western Blotting Markers

Catalog No. 45001182
Change view
Click to view available options
Quantity:
25 Loadings
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
45001182 25 Loadings
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 45001182 Supplier Cytiva Supplier No. RPN810
NLSU: Get 1 Consumable Product When You Buy 1 Obtenez 1 produit consommable à l’achat d’un produit
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Detect proteins on gel and membrane as well as on film

Confirm protein transfer, definite blot orientation and permit calculation of target protein's molecular mass.

  • Prestained markers indicate efficiency of protein transfer to membrane and blot orientation
  • Tagged recombinant protein markers allow accurate calculation of molecular weight of target proteins
  • Compatible with any HRP substrate as well as both PVDF and nitrocellulose membrane types
  • Contain a mixture of three prestained colored markers of defined molecular weight (from 15,000 to 100,000) and seven tagged recombinant proteins (molecular weights from 15,000 to 150,000)

Order Info

Each pack contains ECL DualVue markers (125μL) and S-protein-HRP conjugate (50μL) sufficient for 25 standard gel loadings.

Specifications

Description ECL DualVue Western Blotting Markers contain both colored proteins and tagged proteins that introduce convenience and reliability and facilitate accurate size determination of proteins on Western blots.
Form Liquid
For Use With (Application) Detect proteins on Gel and membrane as well as on film
Kit Contents Pre-Stained Indicator Proteins, Phosphorylase b: 50μg, 100000 (Mr), Rabbit muscle (source), Myoglobin: 50μg, 16000 (Mr), Horse muscle (source), Lysozyme: 50μg, 15000 (Mr), Chicken Egg white (source)
Molecular Weight (g/mol) Mr 15 000
Quantity 25 Loadings
Storage Requirements -20°C
Product Type Protein Molecular Weight Marker