missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Amersham™ ECL Advance™ Blocking Reagent

Catalog No. 45001216
Change view
Click to view available options
Quantity:
40g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
45001216 40g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 45001216 Supplier Cytiva Supplier No. RPN418
Only null left

Use for Controlling background noise. Cytiva Life Sciences™ Amersham™ ECL Advance™ is Blocking Reagent controls background noise while retaining a high level of signal intensity.

Fully compatible with both Hybond™ ECL™ (nitrocellulose) and Hybond-P (PVDF) membranes.

Specifications

Product Line Amersham
For Use With (Application) For blocking Nitrocellulose and PVDF membranes prior to detection with Amersham ECL, Amersham ECL Prime or Amersham ECL Select western blotting detection reagents
Content And Storage 2° to 8°C
Product Type Blocking reagent
Quantity 40g