missing translation for 'onlineSavingsMsg'
Learn More

SOMNI Scientific™ AMD-3 Multiple Delivery Flowmeter Controlled Inhalant Anesthesia Diverter System

Catalog No. 04777102
Change view
Click to view available options
For Use With (Application):
Research Animal Inhalant Anesthesia
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Application)
04777102 Research Animal Inhalant Anesthesia
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 04777102 Supplier SOMNI Scientific™ Supplier No. AMD17004

A flowmeter controlled diverter system that gives the user the ability to deliver accurate flowrates to 3 stations simultaneously. No more guessing about flow. The AMD-3 includes color coded delivery lines making this system very user friendly.

  • 3 Flowmeter Controlled Stations with Toggle Switches
  • Color Coded delivery lines to each station
  • Common Gas outlets for each station
  • Small footprint
  • Adjustable flow rates
  • Stacking design saves valuable workbench space
  • Includes Pressure relief valve and three 6 ft. color coded delivery lines with adapters

Specifications

For Use With (Application) Research Animal Inhalant Anesthesia
For Use With (Equipment) VS-6002, VS-6006, IP-3001, IP-3003, IP-3004, OC-21003, WG-15003, NC-19000
Length (Metric) 30.48 cm
Width (Metric) 20.32 cm
Includes Pressure relief valve and three 6 ft. color coded fresh gas delivery lines with adapters

For Veterinary Use Only