Learn More
Invitrogen™ TAE (10X), RNase-free
Description
Molecular biology grade, Invitrogen 10X TAE is supplied in a 1L bottle. The buffer is certified RNase-free, economical, and ready-to-use. Due to the ubiquitous presence of RNases, manufacturing products for use with RNA is especially challenging. Invitrogen's nuclease-free reagents and buffers are manufactured in facilities specifically designed to prevent the introduction of nucleases. Highly sensitive RNase assays are performed at several different stages of the manufacturing process to ensure the highest quality. These reagents are rigorously tested for contaminating nonspecific endonuclease, exonuclease, and RNase activity. Compatible with agarose gels.
Order Info
Shipping Condition: Room temperature
Specifications
Specifications
| Chemical Name or Material | Running Buffer |
| Concentration | 10X |
| For Use With (Application) | Nucleic acid gel electrophoresis & blotting |
| Physical Form | Liquid |
| Product Line | Ambion |
| Quantity | 1 L |
| Grade | Molecular Biology |
For Research Use Only. Not for use in diagnostic procedures.