missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SSPE (20X), RNase-free

Catalog No. AM9767
Encompass
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AM9767 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AM9767 Supplier Invitrogen™ Supplier No. AM9767

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Molecular biology grade, Invitrogen 20X SSPE solution is supplied in one bottle containing 1 L. The buffer is certified RNase-free, economical, and ready-to-use.

Molecular biology grade, Invitrogen 20X SSPE solution is supplied in one bottle containing 1 L. The buffer is certified RNase-free, economical, and ready-to-use. Due to the ubiquitous presence of RNases, manufacturing products for use with RNA is especially challenging. Invitrogen's nuclease-free reagents and buffers are manufactured in facilities specifically designed to prevent the introduction of nucleases. Highly sensitive RNase assays are performed at several different stages of the manufacturing process to ensure the highest quality. These reagents are rigorously tested for contaminating nonspecific endonuclease, exonuclease, and RNase activity.

Order Info

Shipping Condition: Room temperature

Specifications

Content And Storage Store at room temperature.
Concentration 20 X
Volume (Metric) 1 L
Form Liquid
For Use With (Application) Nucleic Acid Gel Electrophoresis and Blotting
Purity or Quality Grade Molecular Biology Grade
Quantity 1 L
Shipping Condition Room Temperature
Product Line Ambion
Product Type Buffer

For Research Use Only. Not for use in diagnostic procedures.