missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Poly(A) Polymerase (cloned) 2 U/μL

Catalog No. AM2030
Encompass
Change view
Click to view available options
Quantity:
80 units, 2 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AM2030 80 units, 2 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AM2030 Supplier Invitrogen™ Supplier No. AM2030

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Includes

Poly(A) Polymerase, 5X Poly(A) Polymerase Buffer, and 25mM MnCl2

Catalyzes the addition of adenosine to the 3' end of RNA in a sequence-independent fashion

  • The Invitrogen Poly(A) Polymerase enzyme can be used to append radiolabeled ATP to the 3' end of RNA molecules to generate labeled RNAs
  • One tube containing 80U (2U/μL), 5X Poly(A) Polymerase Buffer, and 25mM MnCl2 are provided
  • The enzyme can be used in conjunction with terminal transferase, reverse transcriptase, and thermostable polymerase to clone RNAs of unknown sequence
  • Unit Definition: One unit is the amount of enzyme required to incorporate 1nmol of AMP into acid-insoluble material in 10 minutes at 37°C

DNA & RNA Purification & Analysis, Nucleic Acid Labeling & Oligo Synthesis, PCR & Real-Time PCR, RNA Labeling, Reverse Transcription

Order Info

Shipping Condition: Dry ice

Specifications

Content And Storage Poly(A) Polymerase, 5X Poly(A) Polymerase Buffer, and 25 mM MnCl2 should be stored at –20°C.
Shipping Condition Dry Ice
Polymerase Poly(A) RNA Polymerase
Product Line Ambion
Quantity 80 units, 2 μL
Concentration 5X
Source RNA
Is the Poly(A) Polymerase (cloned) (Cat. No. AM2030) from E. coli?

PolyA polymerase (cloned) is purified from an E. coli strain overexpressing E. coli Poly(A) Polymerase I. Here is the link to the product information sheet: https://assets.thermofisher.com/TFS-Assets/LSG/manuals/4393885_PlyAPlymrseCln_PI.pdf

For Research Use Only. Not for use in diagnostic procedures.