Learn More
Invitrogen™ Nuclease-Free Water (not DEPC-Treated)
Description
Nuclease-free Water (not DEPC-treated) has been deionized, filtered into the final bottle, and autoclaved. It is ready to use and requires no preparation, mixing, or autoclaving. This nuclease-free water is rigorously tested for contaminating nonspecific endonuclease, exonuclease, and RNase activity.
Using Nuclease-free Water
Because this water is not DEPC-treated, it is recommended for applications for which residual DEPC might prove detrimental (e.g., oocyte microinjection).
Order Info
Shipping Condition: Room temperature
Specifications
Specifications
| Grade | Molecular Biology |
| Packaging | Bottle |
| Treatment(s) | Not DEPC-Treated |
| Quantity | 4 x 1000 mL |
| Purification Method | Autoclaved, Membrane-Filtered |
| Product Line | Ambion |
Frequently Asked Questions (FAQs)
Each predesigned TaqMan SNP Genotyping Assay is delivered in a single tube consisting of two differentially labeled allele-specific TaqMan MGB (minor groove binding) probes and a pair of PCR primers. For more information, please see the following link: https://www.thermofisher.com/us/en/home/life-science/pcr/real-time-pcr/real-time-pcr-applications/genetic-variation-analysis-using-real-time/snp-genotyping-with-real-time-pcr.html
Besides the TaqMan SNP Genotyping Assay, you would need to purchase a Genotyping Master Mix and nuclease-free water. A list of compatible master mixes can be found in the following link: https://assets.thermofisher.com/TFS-Assets/LSG/manuals/MAN0009593_TaqManSNP_UG.pdf.
For Research Use Only. Not for use in diagnostic procedures.