missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ MEGAscript™ T7 Transcription Kit

Catalog No. p-4918894
Encompass
Change view
Click to view available options
Product Type:
T7 In Vitro Transcription kit
No. of Reactions:
200 Reactions
25 Reactions
40 Reactions
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type No. of Reactions
AMB13345 T7 In Vitro Transcription kit 200 Reactions
AM1333 T7 In Vitro Transcription kit 25 Reactions
AM1334 T7 In Vitro Transcription kit 40 Reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AMB13345 Supplier Invitrogen™ Supplier No. AMB13345
Only null left

Includes

T7 Enzyme Mix, 10X Reaction Buffer, ATP Solution, CTP Solution, GTP Solution, UTP Solution, pTRI-Xef, TURBO DNase, Ammonium Acetate Stop Solution, Lithium Chloride Precipitation Solution, and Gel Loading Buffer II, Nuclease-free Water

Ultra-high yield technology for in vitro transcription.

The MEGAscript T7 Kit is an ultra-high yield in vitro transcription kit. The high yields are achieved by modifying typical transcription reaction conditions so that very high nucleotide concentrations can be effectively used. The T7 Enzyme Mix and the 10X Reaction Buffer are specifically calibrated for each lot and RNA polymerase.

Using the MEGAscript T7 Kit

The MEGAscript T7 Kit contains in vitro transcription reaction components and a control template. The kit will yield a total of 3–5 mg of RNA (approximately 100 μg of RNA or more per reaction) from the control template supplied with the kit. This corresponds to 400–650 moles of RNA for each mole of template. Smaller templates typically yield a lower mass and a higher molar yield of product. Novel transcription reaction conditions and a patented, high-yield technology allow the synthesis of 10–50 times the amount of RNA produced by conventional transcription reactions.

Applications

The MEGAscript T7 Kit is intended for the synthesis of large amounts of unlabeled or low specific activity RNA for a variety of uses, including in vitro translation, antisense/microinjection studies, and isolation of RNA binding proteins. In large-scale transcription reactions, the concentration of all four nucleotides is high, well above the Km for the enzyme. The MEGAscript T7 Kit typically yields over 10 times more RNA than conventional in vitro transcription reactions. The kit is not recommended for synthesis of high specific activity probes.

Accessory products

MessageAmp aRNA amplification kits are powered by MEGAscript technology and have been developed especially for the amplification of cRNA prior to array analysis. To purify in vitro transcription products from free nucleotides, buffer components, and enzymes, use the MEGAclear™ Kit (Cat. No. AM1908) and MEGAclear-96 Kit (Cat. No. AM1909).

RNA yield: 100 μg

Specifications

Content And Storage T7 Enzyme Mix, 10X Reaction Buffer, ATP Solution, CTP Solution, GTP Solution, UTP Solution, pTRI-Xef, TURBO DNase, Ammonium Acetate Stop Solution, Lithium Chloride Precipitation Solution, and Gel Loading Buffer II are all stored at –20°C. Nuclease-free Water may be stored at any temperature.
Final Product Type Synthetic mRNA, mRNA
Format Kit
Sample Type mRNA, Synthetic mRNA
For Use With (Application) In Vitro Transcription, synthetic mRNA, mRNA, transfection, LNPs, GENEart, linearized DNA plasmid, delivery, PCR products
No. of Reactions 200 Reactions
Product Line Ambion, MEGAscript
Product Type T7 In Vitro Transcription kit
Quantity 200 Reactions
Promoter T7
Shipping Condition Dry Ice
Yield 100 μg
Starting Material PCR Products, Linearized DNA plasmid
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.