missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ MEGAclear™ Transcription Clean-Up Kit

Catalog No. am1908
Encompass
Change view
Click to view available options
Quantity:
20 Preps
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AM1908 20 Preps
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AM1908 Supplier Invitrogen™ Supplier No. AM1908
Only null left

The MEGAclear kit can be used to remove nucleotides, short oligonucleotides, proteins, and salts from RNA. The RNA recovered can be used for any application that requires high purity RNA. Recover from 1 ng to 500 μg of RNA.

The MEGAclear Transcription Clean-up Kit is designed for rapid high-throughput purification of RNA from enzymatic reactions such as in vitro transcription. The kit efficiently separates RNA from unincorporated NTPs, enzymes, and buffer components. The process is simple and fast, taking less than 30 minutes, and it recovers from 1 ng to 500 μg of RNA efficiently. The MEGAclear kit is appropriate for purification of ssRNA larger than 100 nt and dsRNA larger than 200 bp.

The MEGAclear kit purifies transcription reactions to produce high-quality RNA, free of unincorporated NTPs, enzymes, and buffer components, ideal for any downstream application that requires purified RNA. It can be used to remove nucleotides, short oligonucleotides,proteins, and salts from RNA. The RNA recovered can be used for any application that requires high purity RNA.

Use the MEGAclear lit to clean up any of the following:

  • In vitro transcribed RNA:
    • RNA from MEGAscript reactions
    • Amino allyl-modified RNA
    • Biotinyled RNA
    • Cy dye labeled RNA
    • Capped RNA (e.g., from mMESSAGE mMACHINE kit reactions)
  • Total RNA

Using the MEGAclear kit

The protocol is quick and easy. The transcription reaction is mixed with Binding Solution, passed over an RNA-binding filter, and washed; an elution step releases the purified RNA, which is now ready for any application requiring highly pure synthetic or in vitro transcribed RNA. For RNA recovery of 70% or greater, single-stranded RNA should be >100 nt and double-stranded RNA should be >200 bp.

Specifications

Content And Storage Binding Solution, Wash Solution Concentrate, and 5 M Ammonium Acetate are stored at 4°C. Elution Solution can be stored at any temperature, and Filter Cartridges and Collection and Elution Tubes are stored at room temperature.
Final Product Type Purified Synthetic mRNA, Purified mRNA
Format Tube
Isolation Technology Silica Spin Column
Purification Target mRNA, Synthetic mRNA
Sample Type mRNA, Synthetic mRNA
For Use With (Application) In Vitro Transcription, synthetic mRNA, mRNA, transfection, LNPs, GENEart, linearized DNA plasmid, delivery, PCR products
For Use With (Equipment) Vacuum Manifold, Centrifuge
High-throughput Compatibility Not High-throughput Compatible (Manual)
No. of Reactions 20 Preps
Product Line Ambion, MEGAclear
Product Type mRNA purification kit, Purification kit
Quantity 20 Preps
Purification Time 30 min
Shipping Condition Room Temperature
Starting Material Volume 1 ng to 500 μg of dsRNA>200b or ssRA>100bp
Yield 500 μg
Show More Show Less
How do I remove DNase from an RNA sample?

DNase can be removed by the following methods: acid phenol:choloform extraction, lithium chloride precipitation, EDTA, and heat inactivation, or using our MegaClear Transcription Clean-Up Kit (Cat. No. AM1908).

Alternatively, our Invitrogen TURBO DNA-free Kit (Cat. No. AM1907) is supplied with a DNase inactivation reagent that can be used to remove the DNase, as well as divalent cations such as magnesium and calcium, which can catalyze RNA degradation when RNA is heated with the sample.

What is the binding capacity of the MEGAclear kit filter column?

The binding capacity of the 10051G filter columns is 500µg of RNA

Can the Filter Cartridges within the MEGAclear Transcription Clean-Up Kit (Cat. No. AM1908) be ordered separately?

Yes, we do offer the Spin Cartridges (Cat. No. AM10051G) as a stand-alone product, and they would be compatible with the MEGAclear Transcription Clean-Up Kit (Cat. No. AM1908).

Can the collection tubes within the MEGAclear Transcription Clean-Up Kit (Cat. No. AM1908) be ordered separately?

Yes, the Elution Tubes (2.0 mL) (Cat. No. AM12480) are the same that are used in the MEGAclear Transcription Clean-Up Kit (Cat. No. AM1908).

What is the composition of the Elution Solution provided in the MEGAclear Transcription Clean-Up Kit (Cat. No. AM1908)?

It is 0.1 mM EDTA (pH 8.0) in nuclease-free water.

For Research Use Only. Not for use in diagnostic procedures.