missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ MEGAclear™-96 Transcription Clean-Up Kit

Catalog No. am1909
Encompass
Change view
Click to view available options
Quantity:
96 Preps
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AM1909 96 Preps
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AM1909 Supplier Invitrogen™ Supplier No. AM1909
Only null left

Includes

8mL Binding Solution, 20mL Wash Solution Concentrate, 10mL Elution Solution, 20mL Nuclease-free Water, 1 MEGAclear-96 Filter Plate, 1 Collection Plate, 2 Adhesive Plate Sealers

Quickly cleanup large-scale transcription reactions

MEGAclear-96 Kit is designed for quick, high-throughput cleanup of any large-scale transcription reaction. The kit efficiently separates RNA from unincorporated NTPs, enzymes, and buffer components.

  • Purifies transcription reactions in a 96-well format to produce high quality RNA free of unincorporated NTPs, enzymes, and buffer components
  • Quick protocol—purify RNA in less than 20 minutes
  • Can be adapted to robotic platforms
  • Protocol is quick and easy
  • Can also be integrated with your robotic platform or used with manual methods

Specifications

Content And Storage • 8 ml Binding Solution (store at 4°C)
• 20 ml Wash Solution Concentrate (store at 4°C)
• 10 ml Elution Solution (store at room temperature, 4°C, or -20°C)
• 20 ml Nuclease-free Water (store at room temperature, 4°C, or -20°C)
• 1 MEGAclear™-96 Filter Plate (store at room temperature)
• 1 Collection Plate (store at room temperature)
• 2 Adhesive Plate Sealers (store at room temperature)
Final Product Type Purified Synthetic mRNA, Purified mRNA
Format 96-well Plate
Isolation Technology Silica Spin Column
Purification Target mRNA, Synthetic mRNA
Sample Type mRNA, Synthetic mRNA
For Use With (Application) In Vitro Transcription, synthetic mRNA, mRNA, transfection, LNPs, GENEart, linearized DNA plasmid, delivery, PCR products
For Use With (Equipment) Vacuum Manifold, Centrifuge
High-throughput Compatibility High-throughput Compatible
No. of Reactions 96 Preps
Product Line Ambion, MEGAclear
Product Type mRNA purification kit, Purification kit
Quantity 96 Preps
Purification Time 30 min
Shipping Condition Room Temperature
Starting Material Volume 1 ng to 500 μg of dsRNA>200b or ssRA>100bp
Yield 100 μg
Starting Material synthetic mRNA, mRNA
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.