missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ MAXIscript™ T7 Transcription Kit

Catalog No. AM1314M
Encompass
Change view
Click to view available options
Product Type:
T7 In Vitro Transcription Kit
T7 In Vitro Transcription Kit with Manual
Quantity:
100 Reactions
30 Reactions
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type Quantity
AM1314M T7 In Vitro Transcription Kit with Manual 100 Reactions
AM1312 T7 In Vitro Transcription Kit 30 Reactions
AM1314 T7 In Vitro Transcription Kit 100 Reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AM1314M Supplier Invitrogen™ Supplier No. AM1314M

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Includes

T7 Enzyme Mix, 10X Transcription Buffer, ATP Solution, CTP Solution, GTP Solution, UTP Solution, TURBO DNase, Gel Loading Buffer II, pTRI-b-actin, Nuclease-free Water

Fast synthesis of RNA probes for use in ribonuclease protection assays, in situ hybridization, and blot hybridizations

The Ambion™ MAXIscript™ T7 In Vitro Transcription Kit synthesizes RNA probes in just 10 minutes for use in ribonuclease protection assays, in situ hybridization, and blot hybridizations. Each kit contains sufficient reagents for 30 reactions. The MAXIscript In Vitro Transcription Kit synthesizes RNA probes with specific activities reaching 1 × 109 cpm/μg in just 10 minutes. MAXIscript Kits are very efficient at synthesizing full-length probes, even at limiting nucleotide concentrations, making them the perfect companion to ribonuclease protection assays. MAXIscript Kits are also an excellent choice for the synthesis of nonisotopically labeled probes. Labeling can be accomplished post-transcriptionally by using Ambion's BrightStar™ Psoralen-Biotin Labeling Kit, or during transcription by incorporating modified nucleotides into the reaction. MAXIscript Kits can also be used for the synthesis of modest amounts of unlabeled RNA (2–6μg/20μL reaction) of up to 2kb.

  • High yields of high specific activity, full-length RNA probes in just 10 minutes
  • Ideal probe synthesis kit for nuclease protection assays, Northern and Southern blots, and in situ hybridization
  • Includes TURBO DNase™
  • All kits contain quality-tested DNase/RNase-free reagents
  • RNA Yield: 2 to 6μg

Specifications

Content And Storage T7 Enzyme Mix, 10X Transcription Buffer, ATP Solution, CTP Solution, GTP Solution, UTP Solution, TURBO DNase, Gel Loading Buffer II, and pTRI-b-actin are all stored at –20°C. Nuclease-free Water may be stored at any temperature.
Final Product Type Synthetic mRNA, mRNA
Format Kit
Sample Type mRNA, Synthetic mRNA
For Use With (Application) In Vitro Transcription, synthetic mRNA, mRNA, transfection, LNPs, GENEart, linearized DNA plasmid, delivery, PCR products
No. of Reactions 100 Reactions
Product Line Ambion, MAXIscript
Product Type T7 In Vitro Transcription Kit with Manual
Quantity 100 Reactions
Promoter T7
Yield 2-6 μg
Starting Material PCR Products, Linearized DNA plasmid
Show More Show Less
What probe kits do you offer for nonisotopic probe labeling and radiolabeled probe labeling?

We offer two nonisotopic probe labeling kits:

- The BrightStar Psoralen-Biotin Nonisotopic Labeling Kit (Cat. No. AM1480)
- MAXIscript Kits for in vitro transcription (Cat. Nos. AM1308 through AM1326)

We offer three radiolabeled probe labeling kits:

- MAXIscript Kits for in vitro transcription (Cat. Nos. AM1308 through AM1326)
- KinaseMax 5' End-Labeling Kit (Cat. No. AM1520)
- DECAprime II DNA Labeling Kit (Cat. No. AM1455)

What kits should I use to isolate RNA from my sample?

The best isolation kit to use will depend greatly on your application, but, in general, the kits below produce RNA input compatible with the Ion Total RNA-Seq Kit v2 workflow:

Whole Transcriptome (polyA-selected or rRNA-depleted RNA is highly recommended, but total RNA is acceptable)
- Poly A-selected RNA
• Dynabeads mRNA DIRECT Micro Kit (Cat. No. 61021)
• MicroPoly(A)Purist Kit (Cat. No. AM1919)
- rRNA-depleted RNA
• RiboMinus Eukaryote System v2 (Cat. No. A15026)
• Low Input RiboMinus Eukaryote System v2 (Cat. No. A15027)
- Total RNA
• TRI Reagent
• TRizol
• RecoverAll MagMax

For Research Use Only. Not for use in diagnostic procedures.