missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MAXIscript™ SP6 Kit with Manual
Fast synthesis of RNA probes for use in ribonuclease protection assays, in situ hybridization, and blot hybridizations
Specifications
| Content And Storage | SP6 Enzyme Mix, 10X Transcription Buffer, ATP Solution, CTP Solution, GTP Solution, UTP Solution, TURBO DNase, Gel Loading Buffer II, and pTRI-b-actin are all stored at –20°C. Nuclease-free Water may be stored at any temperature. |
|---|---|
| Final Product Type | Synthetic mRNA, mRNA |
| Format | Kit |
| Sample Type | mRNA, Synthetic mRNA |
| For Use With (Application) | In Vitro Transcription, synthetic mRNA, mRNA, transfection, LNPs, GENEart, linearized DNA plasmid, delivery, PCR products |
Includes: SP6 Enzyme Mix, 10X Transcription Buffer, ATP Solution, CTP Solution, GTP Solution, UTP Solution, TURBO DNase, Gel Loading Buffer II, pTRI-b-actin, Nuclease-free Water
| Catalog Number | Mfr. No. | Product Type | Quantity | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Product Type | Quantity | Price | Quantity | ||||
AM1310M
|
Invitrogen™
AM1310M |
SP6 In Vitro Transcription Kit with Manual | 100 Reactions |
N/A
|
|
||||
AM1310
|
Invitrogen™
AM1310 |
SP6 In Vitro Transcription Kit | 100 Reactions |
N/A
|
|
||||
AM1308
|
Invitrogen™
AM1308 |
SP6 In Vitro Transcription Kit | 30 Reactions |
N/A
|
|
||||
AM1308M
|
Invitrogen™
AM1308M |
SP6 In Vitro Transcription Kit with Manual | 30 Reactions |
N/A
|
|
||||
Description
Ambion MAXIscript SP6 In Vitro Transcription Kit synthesizes RNA probes in just 10 minutes for use in ribonuclease protection assays, in situ hybridization, and blot hybridizations. Each kit contains sufficient reagents for 30 reactions.
- High yields of high specific activity, full-length RNA probes in just 10 minutes (RNA Yield: 2-6μg)
- Ideal probe synthesis kit for nuclease protection assays, Northern and Southern blots, and in situ hybridization
- Includes TURBO DNase
Contain quality-tested DNase/RNase-free reagents - Synthesizes RNA probes with specific activities reaching 1 x 109 cpm/μg in just 10 minutes
- Efficient at synthesizing full-length probes, even at limiting nucleotide concentrations, making them perfect companion to ribonuclease protection assays
- For synthesis of nonisotopically labeled probes
- Labeling can be accomplished post-transcriptionally by using Ambion's BrightStar™ Psoralen-Biotin Labeling Kit, or during transcription by incorporating modified nucleotides into reaction
- For synthesis of modest amounts of unlabeled RNA (2–6μg/20μL reaction) of up to 2kb
Specifications
| SP6 Enzyme Mix, 10X Transcription Buffer, ATP Solution, CTP Solution, GTP Solution, UTP Solution, TURBO DNase, Gel Loading Buffer II, and pTRI-b-actin are all stored at –20°C. Nuclease-free Water may be stored at any temperature. | |
| Kit | |
| In Vitro Transcription, synthetic mRNA, mRNA, transfection, LNPs, GENEart, linearized DNA plasmid, delivery, PCR products | |
| SP6 | |
| PCR Products, Linearized DNA plasmid |
| Synthetic mRNA, mRNA | |
| mRNA, Synthetic mRNA | |
| Ambion, MAXIscript | |
| 2-6 μg |
For Research Use Only. Not for use in diagnostic procedures.