missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Gel Loading Buffer II (Denaturing PAGE)

Catalog No. AM8547
Encompass
Change view
Click to view available options
Quantity:
1 mL
10 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AM8547 10 mL
AM8546G 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AM8547 Supplier Invitrogen™ Supplier No. AM8547

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

A 1–2X solution of 95% Formamide, 18 mM EDTA, and 0.025% SDS, Xylene Cyanol, and Bromophenol Blue.

A 1–2X solution of 95% Formamide, 18 mM EDTA, and 0.025% SDS, Xylene Cyanol, and Bromophenol Blue. Supplied in one 1.4 mL bottle. All Invitrogen™ Gel Loading Solutions are rigorously tested for non-specific endonuclease activity, exonuclease activity, RNase activity, and for functionality. These are the same solutions we use in-house and in our kits.

Order Info

Shipping Condition: Room temperature

Specifications

Content And Storage Store at –20°C.
Buffer Sample Loading Buffers
Format Bottle
Label or Dye Bromophenol Blue
Form Liquid
Gel Compatibility Denaturing Polyacrylamide Gels
Quantity 10 mL
Shipping Condition Room Temperature
Product Line Ambion

For Research Use Only. Not for use in diagnostic procedures.