missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ARCA (Anti-Reverse Cap Analog)

Catalog No. AM8045
Encompass
Change view
Click to view available options
Quantity:
10 Units
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AM8045 10 Units
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AM8045 Supplier Invitrogen™ Supplier No. AM8045
Season of Savings
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Includes

Single tube (10 units; 10 A255 U).

Cap analog for in vitro transcription for capped transcripts generation

Invitrogen™ Anti-Reverse Cap Analog (ARCA) is a cap analog used during in vitro transcription for the generation of capped transcripts. A modified cap analog in which the 3' OH group (closer to m7G) is replaced with –OCH3.

  • Tested for contaminating nonspecific endonuclease, exonuclease, and RNase activity
  • Assayed for efficient synthesis of capped in vitro transcripts by using cap analog in functional MEGAscript™ In Vitro transcription and Retic Lysate translation reactions

Specifications

Content And Storage Supplied in a single tube to be stored at or below –70°C. Avoid multiple freeze-thaw cycles. Aliquots of the product may be stored short-term at –20°C.
Concentration 40 mM
Final Product Type Cap Analog for In vitro Transcription, Cap Analog for mRNA
For Use With (Application) In Vitro Transcription, synthetic mRNA, mRNA
For Use With (Equipment) mMessage mMAchine, MEGAscript
Labeling Method RNA Labeling
Product Line Ambion
Product Type Cap analog for mRNA, cap analog for synthetic mRNA
Quantity 10 Units
Shipping Condition Dry Ice

For Research Use Only. Not for use in diagnostic procedures.