missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Supelco™ AmberSep™ (formerly Dowex™) Retardion 11A8, 50-100 mesh

Catalog No. 111015183
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111015183 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111015183 Supplier MilliporeSigma™ Supelco™ Supplier No. 428698500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Amphoteric form

The Retardion 11A8 is not a true mixed bed resin, but a resin with mixed functionality. It contains paired anion and cation exchange sites (each exists as the other's counter-ion) that adsorb mobile ions from a bulk stream.

Specifications

Particle Size 50/100 Mesh
Packaging Poly Bottle
pH 0 to 14
Quantity 500 g
Product Line Ambersep
Capacity (Metric) 0.7 meq/mL by wetted bed volume
MDL Number MFCD00163521
CAS 68955-09-9

Ambersep is a registered trademark of The Dow Chemical Company or an affiliated company of Dow