missing translation for 'onlineSavingsMsg'
Learn More

Ambersep™ 900(OH), ion exchange resin

Catalog No. p-7047367
Change view
Click to view available options
Quantity:
100 g
500 g
1000 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1903522 100 g
AAL1903536 500 g
AAL190350B 1000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL1903522 Supplier Thermo Scientific Chemicals Supplier No. L1903522
Only null left

CAS: 9017-79-2

Specially sized for use in Ambersep resin systems and high-flow rate condensate polishing. It holds regenerable mixed bed applications such as the production of ultrapure water for semiconductor manufacturing.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Specially sized for use in Ambersep resin systems and high-flow rate condensate polishing. It holds regenerable mixed bed applications such as the production of ultrapure water for semiconductor manufacturing.

Solubility
Insoluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Chemical Identifiers

CAS 9017-79-2
MDL Number MFCD00802695

Specifications

Odor Odorless
Quantity 100 g
Solubility Information Insoluble in water.
Chemical Name or Material Ambersep™ 900(OH), ion exchange resin
EINECSNumber 000-000-0
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only