Learn More
Amberlite™ HPR900, OH-form ion-exchange resin
CAS: 197809-12-4
$55.18 - $55.18
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC301341000
|
Thermo Scientific Chemicals
301341000 |
100 g | Plastic bottle |
Each for $55.18
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Ion exchange resinChemical Identifiers
| 197809-12-4 | |
| MFCD00802695 |
Specifications
| Beads | |
| Particle size analysis: uniformity coefficient: <1.45, harmonic mean size: 0.560 to 0.700mm,% <0.300mm: <0.5%,%>1.180mm: <5.0% | |
| volume capacity as is (OH):> 0.9 eq/L | |
| (OH): 66.0 to 75.0% | |
| 100 g |
| 197809-12-4 | |
| Plastic bottle | |
| MFCD00802695 | |
| Ivory | |
| Ambersep™ 900 |
Safety and Handling
GHS P Statement
Causes serious eye damage.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
RUO – Research Use Only