missing translation for 'onlineSavingsMsg'
Learn More

Amberlite™ HPR900, OH-form ion-exchange resin

Catalog No. AC301341000
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC301341000 100 g Plastic bottle
AC301345000 500 g Plastic bottle
AC301340025 2.5 kg Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC301341000 Supplier Thermo Scientific Chemicals Supplier No. 301341000

CAS: 197809-12-4

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Ion exchange resin

Chemical Identifiers

CAS 197809-12-4
MDL Number MFCD00802695

Specifications

Physical Form Beads
Particle Size Particle size analysis: uniformity coefficient: <1.45, harmonic mean size: 0.560 to 0.700mm,% <0.300mm: <0.5%,%>1.180mm: <5.0%
Packaging Plastic bottle
Volume Total Capacity volume capacity as is (OH):> 0.9 eq/L
Water Retention Capacity (OH): 66.0 to 75.0%
Color Ivory
Quantity 100 g
Chemical Name or Material Ambersep™ 900
Hazard Statement GHS P Statement
Causes serious eye damage.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only