missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Supelco™ Ambersep™ 900, Hydroxide Form

Catalog No. 111015848
Change view
Click to view available options
Quantity:
1 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111015848 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111015848 Supplier MilliporeSigma™ Supelco™ Supplier No. 064761KG

Stongly basic (type I) anion exchange resin.

Ambersep™ 900 hydroxide resin has been used for the selective removal of chlorophyll from natural extracts with a high phytochemical content. It may be used as a catalyst for the synthesis of glycerol carbonate by using glycerol and dimethyl carbonate reactants.

Specifications

Quantity 1 kg
Product Line Ambersep
Capacity (Metric) 0.8 meq/mL by wetted bed volume
MDL Number MFCD00802695
CAS 9017-79-2

Ambersep is a registered trademark of The Dow Chemical Company or an affiliated company of Dow