missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ Supelco™ Amberlyst™ A21, Free Base
Description
Use of the solid base is to generate a highly green protocol by eliminating chromatographic purification involving hazardous organic solvents, and for easy recovery and reusability of the catalyst. Amberlyst™ A21 is used as a fluid bed resin. Fluid bed reactor allows gel-type beads, of polymer-supported (PS) organic reactions, to be used in a flow system. The Michael reaction between methyl 1-oxoindan-2-carboxylate and methyl vinyl ketone in toluene can be attained through a fluid bed of Amberlyst™ A21 at 50°C.
Specifications
Specifications
| Packaging | Poly Bottle |
| Quantity | 1 kg |
| Product Line | Amberlyst |
| Capacity (Metric) | 1.3 meq/mL by wetted bed volume |
| MDL Number | MFCD00145842 |
Amberlyst is a registered trademark of The Dow Chemical Company or an affiliated company of Dow