missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ Supelco™ Amberlite™ XAD4
Description
Nonionic macroreticular resin that adsorbs and releases ionic species through hydrophobic and polar interactions; usually used under isocratic conditions.
Specifications
Specifications
| Pore Size | 100 Å |
| Quantity | 500 g |
| Surface Area (Metric) | 750 m2/g |
| Product Line | Amberlite |
| MDL Number | MFCD00132704 |
| Density | 1.08 g/mL (skeletal)(lit.) |
| CAS | 37380-42-0 |
Amberlite is a registered trademark of The Dow Chemical Company or an affiliated company of Dow