missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Supelco™ Amberlite™ XAD4

Catalog No. 111015785
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111015785 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111015785 Supplier MilliporeSigma™ Supelco™ Supplier No. 06444500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Two particle sizes are available: Mesh 20-60 are Cat. Nos. 20276, 10358, XAD4 Mesh 20-50 is Cat. No. 06444

Nonionic macroreticular resin that adsorbs and releases ionic species through hydrophobic and polar interactions; usually used under isocratic conditions.

Specifications

Pore Size 100 Å
Quantity 500 g
Surface Area (Metric) 750 m2/g
Product Line Amberlite
MDL Number MFCD00132704
Density 1.08 g/mL (skeletal)(lit.)
CAS 37380-42-0

Amberlite is a registered trademark of The Dow Chemical Company or an affiliated company of Dow