missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Supelco™ Amberlite™ XAD4, 20-60 mesh

Catalog No. p-7191701
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111016352 100 g
111016349 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111016352 Supplier MilliporeSigma™ Supelco™ Supplier No. SGMA20276
Only null left

Two particle sizes are available: Mesh 20-60 are Cat. Nos. 20276, 10358, XAD4 Mesh 20-50 is Cat. No. 06444

Amberlite™ XAD4, a nonpolar absorbent, used for stripping and concentration of organic compounds. It is also used in developing chelating ligands due to its properties like, porosity, high surface area, purity and durability.

Specifications

Particle Size 20/60 Mesh
Pore Size 100 Å
Quantity 100 g
Surface Area (Metric) 750 m2/g
Product Line Amberlite
MDL Number MFCD00132704
Density 1.08 g/mL (skeletal)(lit.)
CAS 37380-42-0

Amberlite is a registered trademark of The Dow Chemical Company or an affiliated company of Dow