missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Supelco™ Amberlite™ XAD16N, 20-60 mesh

Catalog No. 111015788
Change view
Click to view available options
Quantity:
500 g
1 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111015788 500 g
111015789 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111015788 Supplier MilliporeSigma™ Supelco™ Supplier No. XAD16500G

Hydrophobic polyaromatic resin

Nonionic macroreticular resin that adsorbs and releases ionic species through hydrophobic and polar interactions; usually used under isocratic conditions.

Specifications

Particle Size 20/60 Mesh
Packaging Poly Bottle
Quantity 500 g
Surface Area (Metric) 800 m2/g
Product Line Amberlite
MDL Number MFCD00145831
CAS 104219-63-8

Amberlite is a registered trademark of The Dow Chemical Company or an affiliated company of Dow