missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Supelco™ Amberlite™ XAD1180N, 20-60 mesh

Catalog No. 111015988
Change view
Click to view available options
Quantity:
1 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111015988 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111015988 Supplier MilliporeSigma™ Supelco™ Supplier No. 064741KG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Polymeric adsorbent for hydrophobic compounds: antibiotics, vitamins, amino acids, enzymes; for purification: gallium, myxovirescins, phosphoric acid, riboflavin salts.

Nonionic macroreticular resin that adsorbs and releases ionic species through hydrophobic and polar interactions; usually used under isocratic conditions.

Specifications

Particle Size 20/60 Mesh
Quantity 1 kg
Surface Area (Metric) 450 m2/g
Product Line Amberlite
MDL Number MFCD00145830
CAS 97396-56-0

Amberlite is a registered trademark of The Dow Chemical Company or an affiliated company of Dow