missing translation for 'onlineSavingsMsg'
Learn More

Amberlite XAD-4

Catalog No. AAL1414222
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1414222 100 g
AAL1414236 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1414222 Supplier Thermo Scientific Chemicals Supplier No. L1414222

CAS: 37380-42-0

Polyaromatic adsorbent for small hydrophobic compounds, surfactants, pharmaceutical manufacturing, phenols, chlorinated organics and organic removal from aqueous food streams. It is impregnated with KCN or NaCN, has been used for the conversion of aromatic acyl chlorides to acyl cyanides and of formation of trialkylsilyl chlorides to the silyl cyanides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Polyaromatic adsorbent for small hydrophobic compounds, surfactants, pharmaceutical manufacturing, phenols, chlorinated organics and organic removal from aqueous food streams. It is impregnated with KCN or NaCN, has been used for the conversion of aromatic acyl chlorides to acyl cyanides and of formation of trialkylsilyl chlorides to the silyl cyanides.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agents.

Chemical Identifiers

CAS 37380-42-0
MDL Number MFCD00132704

Specifications

Merck Index 14,382
Quantity 100 g
Solubility Information Insoluble in water.
Chemical Name or Material Amberlite XAD-4
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only