missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Supelco™ Amberlite™ XAD-2

Catalog No. p-7191684
Change view
Click to view available options
Quantity:
100 g
500 g
5 kg
10 kg
25 kg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111016353 100 g
111016350 500 g
111006584 5 kg
111001832 10 kg
111001830 25 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 111016353 Supplier MilliporeSigma™ Supelco™ Supplier No. SGMA20275
Only null left

Amberlite™ XAD™-2 was used: as adsorbent in the extraction of organochlorine compounds using Soxhlat extraction technique, as adsorbent in the extraction of honey samples using column chromatography for HPLC analysis and in adsorption chromatography

Nonionic macroreticular resin that adsorbs and releases ionic species through hydrophobic and polar interactions; usually used under isocratic conditions.

Specifications

Quantity 100 g
Surface Area (Metric) ˜300 m2/g
Product Line Amberlite
Density 1.02 g/mL at 25°C (true wet)(lit.)
CAS 9003-70-7

Amberlite is a registered trademark of The Dow Chemical Company or an affiliated company of Dow