missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ Supelco™ Amberlite™ XAD-2
Description
Nonionic macroreticular resin that adsorbs and releases ionic species through hydrophobic and polar interactions; usually used under isocratic conditions.
Specifications
Specifications
| Quantity | 5 kg |
| Surface Area (Metric) | ˜300 m2/g |
| Product Line | Amberlite |
| Density | 1.02 g/mL at 25°C (true wet)(lit.) |
| CAS | 9003-70-7 |
Amberlite is a registered trademark of The Dow Chemical Company or an affiliated company of Dow