missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Supelco™ Amberlite™ XAD-2

Catalog No. 111001830
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
10 kg
100 g
25 kg
5 kg
500 g
Catalog No. Quantity
111001830 25 kg
111016353 100 g
111016350 500 g
111006584 5 kg
111001832 10 kg
5 options

Catalog No. 111001830

Supplier: MilliporeSigma™ Supelco™ 3025U

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Amberlite™ XAD™-2 was used: as adsorbent in the extraction of organochlorine compounds using Soxhlat extraction technique, as adsorbent in the extraction of honey samples using column chromatography for HPLC analysis and in adsorption chromatography

Nonionic macroreticular resin that adsorbs and releases ionic species through hydrophobic and polar interactions; usually used under isocratic conditions.

Specifications

Quantity 25 kg
Surface Area (Metric) ˜300 m2/g
Product Line Amberlite
Density 1.02 g/mL at 25°C (true wet)(lit.)
CAS 9003-70-7

Amberlite is a registered trademark of The Dow Chemical Company or an affiliated company of Dow