missing translation for 'onlineSavingsMsg'
Learn More

Amberlite XAD-1180

Catalog No. p-7046735
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1609722 100 g
AAL1609736 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1609722 Supplier Thermo Scientific Chemicals Supplier No. L1609722
Only null left

CAS: 97396-56-0

Polymeric adsorbent for hydrophobic compounds: antibiotics, vitamins, amino acids, enzymes; for purification: gallium, myxovirescins, phosphoric acid, riboflavin salts. Amberlite™ XAD1180N can be used as a polymeric adsorbent. The regeneration of this adsorbent has been effective in alkaline medium and tests have shown the adsorbent efficiently removes chromium from tannery waste water. Mainly used in the Amberlite XAD-1180 is a polymeric adsorbent resin that is used primarily in the purification and preparation of hydrophobic compound, Amberlite XAD-1180 is a polymeric adsorbent resin that is used primarily in the purification and preparation of hydrophobic compound, Amberlite XAD-1180 is a polymeric adsorbent resin that is used primarily in the purification and preparation of hydrophobic compound, Amberlite XAD-1180 is a polymeric adsorbent resin that is used primarily in the purification and preparation of hydrophobic compound, Amberlite XAD-1180 is a polymeric adsorbent resin that is used primarily in the purification and preparation of hydrophobic compound.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Polymeric adsorbent for hydrophobic compounds: antibiotics, vitamins, amino acids, enzymes; for purification: gallium, myxovirescins, phosphoric acid, riboflavin salts. Amberlite™ XAD1180N can be used as a polymeric adsorbent. The regeneration of this adsorbent has been effective in alkaline medium and tests have shown the adsorbent efficiently removes chromium from tannery waste water. Mainly used in the Amberlite XAD-1180 is a polymeric adsorbent resin that is used primarily in the purification and preparation of hydrophobic compound, Amberlite XAD-1180 is a polymeric adsorbent resin that is used primarily in the purification and preparation of hydrophobic compound, Amberlite XAD-1180 is a polymeric adsorbent resin that is used primarily in the purification and preparation of hydrophobic compound, Amberlite XAD-1180 is a polymeric adsorbent resin that is used primarily in the purification and preparation of hydrophobic compound, Amberlite XAD-1180 is a polymeric adsorbent resin that is used primarily in the purification and preparation of hydrophobic compound.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place.

Chemical Identifiers

CAS 97396-56-0
MDL Number MFCD00145830

Specifications

Merck Index 14,382
Quantity 100 g
Solubility Information Insoluble in water.
Chemical Name or Material Amberlite XAD-1180
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only