missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ Supelco™ AmberLite MB20 H/OH Mixed Bed Ion Exchange Resin, Hydrogen and Hydroxide Form
Description
Dowex Marathon MR-3 is a premium, premixed, mixed bed product designed for polishing demineralized water to conductivities < 0.1 muS/cm. Marathon MR-3 is a 1:1 equivalent mixture of Marathon C-10 (H) and Marathon A (OH)
Specifications
Specifications
| Packaging | Bottle |
| pH | 0 to 14 |
| Quantity | 100 g |
| Product Line | Marathon |
| Capacity (Metric) | 1.0 meq/mL by wetted bed volume (anion) |
| MDL Number | MFCD00166706 |
Marathon is a trademark of The Dow Chemical Company or an affiliated company of Dow