missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Supelco™ AmberLite MB20 H/OH Mixed Bed Ion Exchange Resin, Hydrogen and Hydroxide Form, 20-50 mesh

Catalog No. 111002508
Change view
Click to view available options
Quantity:
1 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111002508 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111002508 Supplier MilliporeSigma™ Supelco™ Supplier No. 4287361KG

Formerly Dowex™ Marathon™ MR-3 H/OH

Dowex Marathon MR-3 is a premium, premixed, mixed bed product designed for polishing demineralized water to conductivities < 0.1 muS/cm. Marathon MR-3 is a 1:1 equivalent mixture of Marathon C-10 (H) and Marathon A (OH)

Specifications

Particle Size 20/50 Mesh
Packaging Poly Bottle
pH 0 to 14
Quantity 1 kg
Product Line Marathon
Description HCR-S[H+] and SBR[OH-]: in accordance
Capacity (Metric) 1.0 meq/mL by wetted bed volume (anion)
MDL Number MFCD00166706

Marathon is a trademark of The Dow Chemical Company or an affiliated company of Dow