missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ Supelco™ Amberlite™ MB-20, Mixed Bed Ion Exchange Resin, Regenerated
Description
Amberlite™ MB-20 may be used for the removal of ions from aqueous solutions. Uses include primary water chemistry control in once-through systems, ultrapure water production, condensate polishing, in-process and column demineralization, and removal of ionic detergents (such as SDS) from protein samples.
Specifications
Specifications
| Technique | Chromatography |
| Packaging | Poly Bottle |
| Quantity | 250 g |
| Product Line | Amberlite |
| MDL Number | MFCD03791384 |
Amberlite is a registered trademark of The Dow Chemical Company or an affiliated company of Dow